Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Level
species reactivity
vertebrates
manufacturer/tradename
ApoDIRECT, Chemicon®
technique(s)
flow cytometry: suitable
detection method
fluorometric
shipped in
wet ice
General description
Application
Apoptosis & Cancer
Preparation Note
1. The components of this kit are for Research Use only and are not intended for diagnostic procedures.
2. Positive and negative control cells contain 70% (v/v) ethanol as a preservative; wash and reaction buffer contain sodium cacodylate (dimethylarsinic) as a buffer; rinsing and PI/RNase staining buffer contain 0.05% (w/v) sodium azide as a preservative. These materials are harmful if swallowed; avoid areas of contact immediately. See Material Safety Data Sheets.
3. TdT Enzyme will not freeze at -20oC, because it is in 50% (v/v) glycerol solution. Upon warming the TdT enzyme solution, centrifuge the tube for 30 seconds to force all the liquid to the bottom of the tube.
Other Notes
Positive Control Cells,Brown cap
Negative Control Cells, Natural cap
Wash Buffer, Blue cap
Reaction Buffer, Green cap
TdT Enzyme, Yellow cap
F-dUTP, Orange cap
Rinsing Buffer, Red cap
PI/RNase Staining Buffer, Amber bottle
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable | technique(s) flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable |
| detection method fluorometric | detection method fluorometric | detection method fluorometric | detection method fluorometric |
| shipped in wet ice | shipped in wet ice | shipped in dry ice | shipped in dry ice |
| manufacturer/tradename ApoDIRECT, Chemicon® | manufacturer/tradename - | manufacturer/tradename ApopTag, Chemicon® | manufacturer/tradename ApopTag, Chemicon® |
| species reactivity vertebrates | species reactivity - | species reactivity - | species reactivity - |
| Quality Level 100 | Quality Level 300 | Quality Level 100 | Quality Level 100 |
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1
Storage Class
3 - Flammable liquids
flash_point_f
69.8 °F
flash_point_c
21 °C
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| APT110 | 04053252001000 |




