Skip to Content
Merck

Skip To

5.89591

1,4-Dioxane

anhydrous, 99.8%

Synonym(s):

Diethylene oxide, Dioxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
NACRES:
NA.22
UNSPSC Code:
12352005
Beilstein/REAXYS Number:
102551
MDL number:
Assay:
99.8%
Grade:
anhydrous
Bp:
100-102 °C (lit.)
Vapor pressure:
27 mmHg ( 20 °C), 40 mmHg ( 25 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

Quality Segment

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C), 40 mmHg ( 25 °C)

assay

99.8%

autoignition temp.

356 °F

expl. lim.

22 %

refractive index

n20/D 1.422 (lit.)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 1,4-Dioxane


signalword

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

supp_hazards



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
589591010004065267986609
589591025004065267986616
589591100004065267986623
589591200004065267986630

Questions

  1. ¿tiene tapón septum?

    1 answer
    1. The type of top for this product depends on the package size ordered.

      Helpful?

Reviews

No rating value

Active Filters