Product Name
Diethyl pyrocarbonate 96% (NT)
Product Number
D5758
Product Brand
SIGMA
CAS Number
1609-47-8
Molecular Weight
162.14
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (NT) HCLO4 0.1M≥ 95.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE, ≥ 97 %
RECOMMENDED RETEST PERIOD24 MONTHS