Merck
Skip To
MABS2140
Anti-Active HGF Antibody, clone t8E4
clone t8e4
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
t8e4, monoclonal
Species reactivity:
-
Application:
Citations:
-
Let Us Assist
clone
t8e4, monoclonal
Quality Level
isotype
IgG1κ
target post-translational modification
unmodified
Immunogen
His-tagged recombinant fragment corresponding to K2SP portion of human hepatocyte growth factor (HGF).
Biochem/physiol Actions
Clone tc8E4 is a mouse monoclonal antibody that detects the active form of human hepatocyte growth factor (HCF). It targets an epitope within the K4 and SP domains in the C-terminal half.
Preparation Note
1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at +2°C to +8°C from date of receipt.
Disclaimer
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
MABE971MABT171MABT826
clone
t8e4, monoclonal
clone
12A5.1, monoclonal
clone
4A3, monoclonal
clone
2G2, monoclonal
isotype
IgG1κ
isotype
IgG2aκ
isotype
IgG1κ
isotype
IgMκ
Quality Level
200
Quality Level
100
Quality Level
100
Quality Level
100
target post-translational modification
unmodified
target post-translational modification
unmodified
target post-translational modification
unmodified
target post-translational modification
unmodified
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Visit the Document Library
Global Trade Item Number
SKUGTIN
MABS2140-100UG04065265085885
MABS2140-25UG04065265085892