ホーム透明導電性基板 (ITO・FTO)
透明導電性基板 (ITO・FTO)
FTO透明導電性ガラス
FTO(Fluorine-doped tin oxide)透明導電性ガラスは、オプトエレクトロニクスやタッチスクリーン、薄膜太陽電池、省エネ窓、RFI/EMI遮蔽をはじめとするさまざまな用途に理想的な材料です。フッ素ドープ酸化スズは、大気条件下で比較的安定であり、化学的に安定で機械強度が高く、さらに、高温耐熱性をもち、物理的剥離にも強く、酸化インジウムスズよりも安価なため、非常に有望な材料です。
TEC Glass™は、コストパフォーマンスに優れていることで知られています。さらに、FTOを用いた基板は、TiO2やZnOナノ構造薄膜へのインジウムの拡散の問題を避けることができ、熱特性や光透過性に優れています。
製品名 | サイズ(L x W x D) | 透過率(%) | 表面抵抗(Ω/sq) | 製品番号 |
---|---|---|---|---|
Fluorine doped tin oxide coated glass slide, FTO, TEC 7 | 50 × 50 × 2.2 mm | 80-82 | 7 | 735140-5EA |
100 × 100 × 2.3 mm | 80-82 | 7 | 735159-5EA | |
300 × 300 × 2.2 mm | 80-82 | 7 | 735167-1EA | |
Fluorine doped tin oxide coated glass slide, FTO, TEC 8 | 50 × 50 × 3 mm | 80-82 | 8 | 735175-5EA |
100 × 100 × 3 mm | 80-82 | 8 | 735183-5EA | |
300 × 300 × 3.2 mm | 80-82 | 8 | 735191-1EA | |
Fluorine doped tin oxide coated glass slide, FTO, TEC 10 | 50 × 50 × 3 mm | 83 | 10 | 735205-5EA |
100 × 100 × 3 mm | 83 | 10 | 735213-5EA | |
300 × 300 × 3 mm | 83 | 10 | 735221-1EA | |
Fluorine doped tin oxide coated glass slide, FTO, TEC 15 | 50 × 50 × 2.2 mm | 82-85 | 13 | 735248-5EA |
100 × 100 × 2 mm | 82-85 | 13 | 735256-5EA | |
300 × 300 × 2.3 mm | 82-85 | 13 | 735264-1EA |
ITO透明導電性基板
PEN基板
製品名 | サイズ(L x W x D) | 表面抵抗(Ω/sq) | 透過率(%) | 製品番号 |
---|---|---|---|---|
Indium tin oxide coated PEN with titanium(IV) oxide buffer layer | 29.7 cm × 21 cm × 125 μm | <15 | 80 | 918067-1EA |
Indium tin oxide coated PEN | 80 | 917826-1EA | ||
Indium tin oxide coated PEN, with 0% transparent Pt | 9.6 cm × 9.6 cm × 125 μm | 5 | 0 | 917311-1EA |
Indium tin oxide coated PEN, with 60% transparent Pt | 60 | 917567-1EA |
PET基板
製品名 | サイズ(L x W x D) | ITO膜厚 (Å) | 表面抵抗(Ω/sq) | 透過率(%) | 製品番号 |
---|---|---|---|---|---|
Indium oxide coated PET, IO-PET, Slide | 150 x 150 x 0.2 mm | - | ≦ 10 | - | 700177-5PAK, 10PAK |
Indium tin oxide coated PET, ITO-PET, Sheet 1 | 1 ft x 1 ft x 5 mil | 1,000 | 60 | >79 | 639303-1EA, 5EA ※1 |
750 | 100 | >77 | 639281-1EA, 5EA ※1 | ||
1 ft x 1 ft x 7 mil | 1,000 | 60 | - | 749729-1EA, 5EA | |
1 ft x 1 ft x 7 mil | 1,000 | 100 | - | 749737-1EA, 5EA | |
1 ft x 1 ft x 5 mil | 1,000 | 200 | - | 749745-1EA, 5EA | |
1 ft x 1 ft x 7 mil | 1,000 | 200 | - | 749753-1EA, 5EA | |
1 ft x 1 ft x 5 mil | 1,000 | 250 | - | 749761-1EA, 5EA | |
1 ft x 1 ft x 7 mil | 1,000 | 250 | - | 749788-1EA, 5EA | |
1 ft x 1 ft x 5 mil | 1,000 | 300 | - | 749796-1EA, 5EA | |
1 ft x 1 ft x 7 mil | 1,000 | 300 | - | 749818-1EA, 5EA |
ガラス基板
製品名 | サイズ(L x W x D) | ITO膜厚 (Å) | 表面抵抗(Ω/sq) | 透過率(%) | 製品番号 |
---|---|---|---|---|---|
Indium tin oxide coated glass slide, rectangular, ITO 1 | 75 x 25 x 1.1 mm | 1,200-1,600 | 8 – 12 | >83 | 578274-10PAK, 25PAK |
600-1,000 | 15 – 25 | >78 | 636916-10PAK, 25PAK | ||
300-600 | 30 – 60 | >84 | 636908-10PAK, 25PAK | ||
150-300 | 70 – 100 | >87 | 576352-10PAK, 25PAK | ||
Indium tin oxide coated glass slide, square, ITO 2 | 25 x 25 x 1.1 mm | 1,200-1,600 | 8 – 12 | ~84 | 703192-10PAK |
300-600 | 30 – 60 | ~84 | 703184-10PAK | ||
150-300 | 70 – 100 | ~86 | 703176-10PAK | ||
Indium tin oxide coated aluminosilicate glass slide 3 | 75 x 25 x 1.1 mm | 1,200-1,600 | 5 – 15 | >85 | 576360-10PAK, 25PAK |
関連情報