Merck
DWK73760-16125
KIMBLE® disposable screw thread culture tube
flat bottom borosilicate glass, tube diam. × L (16 mm) (125 mm), non-sterile
About This Item
UNSPSC kód:
41121706
NACRES:
NB.11
material
flat bottom borosilicate glass
flat bottom
screw top neck
transparent borosilicate glass tube
sterilitás
non-sterile
kiszerelés
case of 1000 ea Modular tray shrink-wrapped in plastic film
gyártó/kereskedő neve
Kimble® 73760-16125
cső átmérő × H
(16 mm) (125 mm)
szerelvény
neck (External Thread)
Általános leírás
Disposable Screw Thread Culture Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Tulajdonságok és előnyök
• Manufactured from borosilicate glass 5.1• 73760 series and 90 series items that end in ‘F′ have flat bottoms which contribute to stability• 73770 series items are produced with a round bottom• Modular trays are shrink-wrapped in plastic film• Supplied without caps• No marking spot
Jogi információk
KIMBLE is a registered trademark of DWK Life Sciences
Válasszon a legfrissebb verziók közül:
Analitikai tanúsítványok (COA)