Merck
Z675601
Polyethylene carboy with spigot
size 20 L
Größe auswählen
1 EA
€ 277,00
€ 277,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 277,00
About This Item
UNSPSC-Code:
41121820
NACRES:
NB.35
€ 277,00
Check Cart for Availability
Materialien
high-density polyethylene (Bottle and spigot)
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F11847-0050
Größe
20 L
Allgemeine Beschreibung
These bottles and carboys have a screw in dispensing spigot that can be replaced with a plug (included) when the vessels are used for long-term storage. Heavy knurling on the cap provides excellent grip for opening and closing. The smooth sides accept adhesive labels and can be marked with most indelible markers.
  • Spigot opens fully in one half-turn
  • Molded of polyethylene, which resists corrosive acids and alkalis
  • Extra heavy walls, yet lighter than glass
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)