Skip to Content
Merck
All Photos(1)

Documents

1.02781

Supelco

2-Propanol

≥99.9% (GC), suitable for LC/MS, suitable for ASTM® 7979, EPA 8327, ASTM® 7968, LiChrosolv®

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
51471904
EC Index Number:
200-661-7
NACRES:
NA.03

product name

2-Propanol, hypergrade for LC-MS LiChrosolv®

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 8327

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

LC/MS: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water
≤10 ppb Al (Aluminium)
≤10 ppb Ca (Calcium)
≤10 ppb Fe (Iron)
≤10 ppb Mg (Magnesium)
≤150 ppb Na (Sodium)
≤5 ppb Any further metal (ICP-MS)
≤5 ppb K (Potassium)

evapn. residue

≤2.0 mg/L

color

APHA: ≤10

transmittance

220 nm, ≥80% (Checking after purging with N2)
230 nm, ≥90% (Checking after purging with N2)
250 nm, ≥99% (Checking after purging with N2)

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Color: ≤ 10 Hazen
Water: ≤ 0.05 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Gradient grade (at 235 nm): ≤ 1.0 mAU
Gradient grade (at 254 nm): ≤ 1.0 mAU
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 0.5 ppb
Transmission (Checking after purging with N₂ (at 220 nm)): ≥ 80 %
Transmission (Checking after purging with N₂ (at 230 nm)): ≥ 90 %
Transmission (Checking after purging with N₂ (from 250 nm)): ≥ 99 %
Al (Aluminium): ≤ 10 ppb
Ca (Calcium): ≤ 10 ppb
Fe (Iron): ≤ 10 ppb
K (Potassium): ≤ 5 ppb
Mg (Magnesium): ≤ 10 ppb
Na (Sodium): ≤ 150 ppb
Any further metal (ICP-MS): ≤ 5 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 10 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI negative): ≤ 20 ppb
Filtered by 0.2 µm filter.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service