Skip to Content
Merck

Skip To

PP0100

Trypsin Profile IGD Kit

Synonym(s):

In gel digestion kit, Proteomics grade Trypsin, TPCK treated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

 kit sufficient for ≤100 applications

Quality Level

storage temp.

2-8°C

General description

The Trypsin Profile IGD Kit enables fast, efficient, and complete in-gel tryptic digestion of up to 100 excised protein spots. Digested proteins are ready for MALDI-MS (matrix-assisted laser desorption/ionization-mass spectrometry) and require no additional preparation. Because the Trypsin Profile IGD Kit contains Proteomics Grade Trypsin, a higher sequence coverage and fewer ambiguous autolytic peaks are observed in MALDI spectra.
Trypsin has been chemically modified through reductive methylation of the ε-amino groups of lysine to reduce autolysis and minimize autolytic fragments. In addition, it has been TPCK (tosyl phenylalanyl chloromethyl ketone) treated to remove residual chymotrypsin activity and then further purified by affinity chromatography, yielding a highly purified trypsin suitable for proteomics work.

Application

Trypsin Profile IGD Kit has been used for in-gel digestion for LC-MS (liquid chromatography-mass spectrometry)/MS and MALDI-TOF (matrix assisted laser desorption ionization-time of flight) mass spectrometry analysis.[1]

Features and Benefits

  • Faster destaining than with alternative in-gel digest kits
  • Includes 100% of the reagents needed to destain, digest, and extract proteins/peptides of interest
  • Compatible with Coomassie, SYPRO Orange, SYPRO Ruby, and ProteoSilver Stained gels
  • Resulting samples are ready for analysis by MALDI-MS or HPLC-MS

Legal Information

ProteoSilver is a trademark of Sigma-Aldrich Co. LLC
SYPRO is a trademark of Thermo-Fisher Scientific, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T7575TT0010EMS0005
usage

 kit sufficient for ≤100 applications

usage

-

usage

-

usage

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Kit Components Only

Product No.
Description

  • Destaining Solution Reconstituted to 75

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

We have developed a novel, high-binding capacity, antibodybased resin for the depletion of twenty high abundance proteins in human plasma. The technology is a significant improvement over existing kits that remove 2, 6, or 12 proteins.

View All Related Content

Identification of plant viruses using one-dimensional gel electrophoresis and peptide mass fingerprints.
Luo, H., et al.
J. Virol. Methods, 165(2), 297-301 (2010)
SILAM for quantitative proteomics of liver Akt1/PKBa after burn injury.
Lu XM, et al.
International Journal of Molecular Medicine, 29, 461-461 (2012)
X-M Lu et al.
International journal of burns and trauma, 3(1), 37-48 (2013-02-07)
Insulin resistance is a major effect of burn injury and insulin receptor substrate-1 (IRS-1) plays an important role in signal transduction. Here, we explored the integrity of IRS-1 in muscle after burn injury. A murine model of severe burn injury



Global Trade Item Number

SKUGTIN
PP0100-1KT04061835574728

Questions

Reviews

No rating value

Active Filters