Skip to Content
Merck
All Photos(1)

Key Documents

650501

Sigma-Aldrich

Acetone

Acetone

HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€70.70

€70.70


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 L
€70.70

About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12190000
eCl@ss:
39021201
PubChem Substance ID:
NACRES:
NA.02
grade:
HPLC Plus
HPLC grade
for residue analysis
Assay:
≥99.9%
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

€70.70


In StockDetails


Request a Bulk Order

grade

HPLC Plus
HPLC grade
for residue analysis

Quality Level

Agency

suitable for EPA 1613
suitable for EPA 1633

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

expl. lim.

13.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water
≤1.0 ppb Fluorescence (quinine) at 365 nm

evapn. residue

≤0.0003%

halogenated residue

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

solubility

water: miscible

density

0.791 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 330 nm Amax: 1.0
λ: 340 nm Amax: 0.06
λ: 350 nm Amax: 0.01
λ: 375-400 nm Amax: 0.005

application(s)

food and beverages

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is a ketone. Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700°C or 650°C has been reported to efficiently photocatalyze oxidation of acetone.[1] Its aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions.[2] It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide.[3] Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.[4]

Application

Acetone may be used in the following studies:
  • NMR spectroscopic evaluation of glucose metabolism in biological samples (blood).[5]
  • Preparation of 13wt% amorphous poly (lactic acid) (PLA) solution, which was required for the preparation of electrospun PLA membranes.[6]
  • As a precursor for the synthesis of methyl isobutyl ketone (MIBK) in the presence of sulfonated graphene oxide-Pd/cordierite catalyst.[7]
  • Synthesis of (4-hydroxymethyl-2,2-dimethyl-1,3-dioxolane), a solketal from glycerol using supercritical fluids (SCF) technology.[8]
It may be used in the synthesis of the following acetone hydrazones:
  • 1-isopropylidene-2-methylhydrazine[9]
  • 1-isopropylidene-2-hydroxyethylhydrazine[9]
  • 1-isopropylidene-2-formylhydrazine[9]
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
Suitable for HPLC, spectrophotometry, environmental testing

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F

Flash Point(C)

-21.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sulfonated graphene oxide supported Pd bifunctional catalyst for one-pot synthesis of methyl isobutyl ketone from acetone with high conversion and selectivity.
Liu M, et al.
J. Mol. Catal. A: Chem., 408, 85-90 (2015)
Development of eco-efficient micro-porous membranes via electrospinning and annealing of poly (lactic acid).
Li L, et al.
Journal of Membrane Science, 436, 57-67 (2013)
Saskia Kley et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 296(4), R936-R943 (2009-02-06)
Obesity is a risk factor for type 2 diabetes in cats. The risk of developing diabetes is severalfold greater for male cats than for females, even after having been neutered early in life. The purpose of this study was to
Synthesis and characterization of acetone hydrazones.
Miro SC and Delalu H.
Zeitschrift fur Anorganische und Allgemeine Chemie, 638(1), 57-63 (2012)
Ketalization of glycerol to solketal in supercritical acetone.
Royon D, et al.
Journal of Supercritical Fluids, 58(1), 88-92 (2011)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. quel est origine de l'acétone HPLC? synthèse, biosourcing?

    1 answer
    1. This product is typically manufactured as synthetic organic, however, may vary lot to lot. This information can be found on the lot specific Certificate of Origin. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a Certificate under the ‘Certificate of Origin’ tab:
      https://www.sigmaaldrich.com/product/sigald/650501#product-documentation

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service