Skip to Content
Merck
All Photos(1)

Documents

SML0148

Sigma-Aldrich

Imidapril hydrochloride

≥98% (HPLC)

Synonym(s):

(4S)-3-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1-methyl-2-oxo-4-imidazolidinecarboxylic acid hydrochloride, Novaloc, TA 6366, Tanapril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27N3O6 · HCl
CAS Number:
Molecular Weight:
441.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -50 to -70° in ethanol (c=0.5)

color

white to tan

solubility

H2O: ≥5 mg/mL

originator

Trinity Pharma Solutions

storage temp.

−20°C

SMILES string

Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@@H](CN(C)C2=O)C(O)=O

InChI

1S/C20H27N3O6.ClH/c1-4-29-19(27)15(11-10-14-8-6-5-7-9-14)21-13(2)17(24)23-16(18(25)26)12-22(3)20(23)28;/h5-9,13,15-16,21H,4,10-12H2,1-3H3,(H,25,26);1H/t13-,15-,16-;/m0./s1

InChI key

LSLQGMMMRMDXHN-GEUPQXMHSA-N

General description

Imidapril comprises large acyl moiety and is hydrolyzed by carboxylesterase (CES) 1.

Application

Imidapril hydrochloride may be used to test its effect on pharyngeal and laryngeal muscle to treat impaired swallowing.
Imidapril hydrochloride was used as a standard in bioequivalence test by LC/MS method.

Biochem/physiol Actions

Imidapril hydrochloride is a long-acting inhibitor of angiotensin converting enzyme used in the treatment of hypertension, congestive heart failure and diabetic nephropathy. It restores decreased airway sensation and bladder sensation in patients with multiple sclerosis.
Imidapril is a potent angiotensin converting enzyme inhibitor and anti-hypertensive.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Trinity Pharma Solutions. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fen Wei et al.
Heart (British Cardiac Society), 97(6), 479-484 (2011-02-08)
Anti-angiotensin II receptor subtype 1 (AT1 receptor) autoantibodies have previously been shown in sera of hypertensive patients. This study assessed whether anti-AT1-receptor autoantibody in serum is correlated with the efficacy of an AT1-receptor blocker (ARB; candesartan)-based regimen in hypertensive patients
Sayaka Nagata et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 35(2), 234-238 (2011-10-14)
It has been suggested that proangiotensin-12 (proang-12), a novel angiotensin peptide recently discovered in rat tissues, may function as a component of the tissue renin-angiotensin system (RAS). To investigate the role of proang-12 in the production of angiotensin II (Ang
Ji Hye Yun et al.
Archives of pharmacal research, 28(4), 463-468 (2005-05-28)
The purpose of the present study was to develop a standard protocol for imidapril hydrochloride bioequivalence testing. For this reason, a specific LC-MS method was developed and validated for the determination of imidapril in human plasma. A solid-phase extraction cartridge
Yi-Fei Dong et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(9), 2911-2920 (2011-05-20)
The purpose of this work was to test whether brain-penetrating angiotensin-converting enzyme (ACE) inhibitors (e.g., perindopril), as opposed to non-brain-penetrating ACE inhibitors (e.g., enalapril and imidapril), may reduce the cognitive decline and brain injury in Alzheimer's disease (AD). We first
Mitsuru Ohishi et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 33(11), 1150-1154 (2010-08-13)
The principal means for reducing proteinuria in patients with chronic kidney disease are strong blockade of the renin-angiotensin system and strict regulation of blood pressure (BP). This study compared the efficacy of the maximum permissible doses of two common angiotensin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service