Skip to Content
Merck
All Photos(1)

Key Documents

1082300

USP

Butoconazole nitrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17Cl3N2S · HNO3
CAS Number:
Molecular Weight:
474.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

butoconazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC1=C(C(Cl)=CC=C1)SC(CCC2=CC=C(Cl)C=C2)CN3C=CN=C3.O[N+]([O-])=O

InChI

1S/C19H17Cl3N2S.HNO3/c20-15-7-4-14(5-8-15)6-9-16(12-24-11-10-23-13-24)25-19-17(21)2-1-3-18(19)22;2-1(3)4/h1-5,7-8,10-11,13,16H,6,9,12H2;(H,2,3,4)

InChI key

ZHPWRQIPPNZNML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butoconazole nitrate (BN) is a imidazole used as an antifungal agent.

Application

Butoconazole nitrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Butoconazole Nitrate Vaginal Cream.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-meng Jia et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 34(3), 431-436 (2014-06-19)
A liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the determination of butoconazole in human plasma. Human plasma samples of 0.2 μL were pretreated by a single step protein precipitation procedure and analyzed using a high performance
J B Jacobson et al.
Acta obstetricia et gynecologica Scandinavica, 64(3), 241-244 (1985-01-01)
The continuously increasing incidence of vulvovaginal candidiasis necessitated a search for novel therapeutic modalities. Butoconazole nitrate (BN) a new imidazole, has been singled out for clinical studies since, in experimental vaginal candidiasis, it proved more effective than either miconazole nitrate
[Butoconazole nitrate in vulvovaginal candidiasis. A comparative study with clotrimazole].
E Herrera Nava et al.
Ginecologia y obstetricia de Mexico, 56, 138-143 (1988-05-01)
[Action of 2 new imidazole derivatives (butoconazole and sulconazole) on Candida, Trichophyton and Aspergillus].
J P Latge et al.
Annales de dermatologie et de venereologie, 116 Suppl 1, 20-23 (1989-01-01)
H Ruf et al.
The Journal of international medical research, 18(5), 389-399 (1990-09-01)
In a randomized, single-blind, parallel study the safety and efficacy of 2% butoconazole nitrate cream used for 3 days were compared with those of 1% econazole nitrate cream used for 7 days at night in patients with vulvovaginal candidiasis. Patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service