Skip to Content
Merck

Skip To

UPS1

Universal Proteomics Standard Set

Protein Mass Spectrometry Calibration Standard

Synonym(s):

Standard Set

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12161503
EC Number:
232-650-8
NACRES:
NA.24
Form:
ready-to-use solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

ready-to-use solution

Quality Level

quality

Protein Mass Spectrometry Calibration Standard

technique(s)

mass spectrometry (MS): suitable

storage temp.

−20°C

General description

The Universal Proteomics Standard (UPS) Set was developed in collaboration with the Association of Biomolecular Resource Facilities (ABRF) Proteomics Standards Research Group (sPRG). This protein mixture was extensively evaluated and reported under the direction of ABRF′s sPRG during a comprehensive 2005/2006 study. The findings of the study were presented at the ABRF 2006 and US HUPO 2006 conferences.

Application

The Universal Proteomics Standard (UPS) Set is intended to standardize and/or evaluate mass spectrometric (e.g., LC-MS/MS, MALDI-TOF-MS, etc.) and electrophoretic analysis conditions prior to the analysis of complex protein samples.[1][2][3][4][5][6] Potential uses include:
  • Bracketing critical experimental datasets for confirming the robustness of analysis methods
  • Comparison of MS or other proteomic data that are generated in different labs using a variety of analytical strategies and instruments
  • Identifying limitations of proteomics analysis systems and search algorithms
  • An external reference to assist with the evaluation of data derived from poorly defined samples

Features and Benefits

Discover the Benefits for Yourself!
  • Test the power of your analytical strategy
  • Troubleshoot and optimize your analytical protocol
  • Confirm system suitability before analyzing critical samples
  • Normalize analytical results day to day or lab to lab

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UPS2EMS0004MSST0016
technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

-

technique(s)

mass spectrometry (MS): suitable

form

ready-to-use solution

form

ready-to-use solution

form

ready-to-use solution

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

quality

Protein Mass Spectrometry Calibration Standard

quality

Protein Mass Spectrometry Calibration Standard

quality

-

quality

-

Proteomics Standards, Proteomics Controls, Proteomics Standardization, Proteomics standard, ProteoNorm, ProteoStandard, Proteomics, proteomics standard kit, mass spectrotmetry, LC-MS, MALDI-TOF, electrophoresis, dynamic range, orbi-trap, lcms, UPS1, hypergeometric, quadrupole, Proteomics Standards, Proteomics Controls, Proteomics Standardization, Proteomics standard, Pr

Proteomics Standards, Proteomics Controls, Proteomics Standardization, Proteomics standard, ProteoNorm, ProteoStandard, Proteomics, mass spectrotmetry, LC-MS, MALDI-TOF, electrophoresis, dynamic range, orbi-trap, lcms, UPS2, hypergeometric, quadrupole, Proteomics Controls, ProteoNorm,

-

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated 20 μg
    SDS

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Matthew The et al.
Nature communications, 11(1), 3234-3234 (2020-06-28)
In shotgun proteomics, the analysis of label-free quantification experiments is typically limited by the identification rate and the noise level in the quantitative data. This generally causes a low sensitivity in differential expression analysis. Here, we propose a quantification-first approach
Christian J Koehler et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 25(1), 61-66 (2019-11-02)
Proteolytic digestion prior to LC-MS analysis is a key step for the identification of proteins. Digestion of proteins is typically performed with trypsin, but certain proteins or important protein sequence regions might be missed using this endoproteinase. Only few alternative
Amanda G Paulovich et al.
Molecular & cellular proteomics : MCP, 9(2), 242-254 (2009-10-28)
Optimal performance of LC-MS/MS platforms is critical to generating high quality proteomics data. Although individual laboratories have developed quality control samples, there is no widely available performance standard of biological complexity (and associated reference data sets) for benchmarking of platform



Global Trade Item Number

SKUGTIN
UPS1-5X1KT04061835575084
UPS1-1KT04061835522682

Questions

Reviews

No rating value

Active Filters