Skip to Content
Merck
All Photos(2)

Key Documents

1.07288

Supelco

n-Pentane

hypergrade for organic trace analysis SupraSolv®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
35-36 °C (lit.)

vapor density

2.48 (vs air)

Quality Level

product line

SupraSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

technique(s)

gas chromatography (GC): suitable

impurities

≤0.01% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® hypergrade for organic trace analysis is a unique and universal solvent quality which can be employed for all three GC detection methods (ECD, FID and MS). Thanks to the large retention time range, even low-boiling substances can be reliably analyzed.

Application

The solvent is featured in studies such as:
  • quantification of trace odor-active polyfunctional mercaptans in wine by GC-MS
  • analysis of volatiles from Brussels sprouts plants infested by herbivorous insects by GC coupled to electroantennogram (EAG)
  • analysis of herbicides molinate and bentazone in soil by GC coupled to flame ionization detector(FID)

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 3.0 mg/l
Water: ≤ 0.01 %
Colour: ≤ 10 Hazen
GC/ECD (retention range 1,2,4-tri-chlorobenzene to decachlorobiphenyle,individual signals (lindane standard)): ≤ 2 pg/ml
GC/ECD (retention range dichloromethane to 1,2,4-trichlorobenzeneindividual signals (tetrachloromethane)): ≤ 1 ng/ml
GC/FID (retention range n-undecane ton-tetracontane,individual signals (n-tetradecane standard)): ≤ 2 ng/ml
GC/MSD (retention range n-undecane ton-tetracontane, scaning area 30 - 600 amu,individual signals (n- tetradecane standard)): ≤ 2 ng/ml
suitable for residue analysis

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of organic contaminants from soils by an electrokinetic process: the case of molinate and bentazone. Experimental and modeling
Ribeiro A, et al.
Separation & Purification Reviews, 79(2), 193-203 (2011)
Selective preconcentration of volatile mercaptans in small SPE cartridges: Quantitative determination of trace odor-active polyfunctional mercaptans in wine
Mateo-Vivaracho L, et al.
Journal of Separation Science, 32(21), 3845-3853 (2009)
GC-EAG-analysis of volatiles from Brussels sprouts plants damaged by two species of Pieris caterpillars: olfactory receptive range of a specialist and a generalist parasitoid wasp species
Smid HM, et al.
Chemoecology, 12(4), 169-176 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service