Skip to Content
Merck
All Photos(1)

Key Documents

1.03701

Supelco

Hexane

≥99% (GC), suitable for LC/MS, LiChrosolv®

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
41116105
EC Index Number:
203-777-6
NACRES:
NA.03
technique(s):
HPLC: suitable
LC/MS: suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

n-Hexane, hypergrade for LC-MS LiChrosolv®

vapor density

~3 (vs air)

Quality Level

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

LiChrosolv®

Assay

≥99% (GC)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable
LC/MS: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water
≤10 ppb Al (Aluminium)
≤10 ppb Ca (Calcium)
≤10 ppb Fe (Iron)
≤10 ppb Mg (Magnesium)
≤10 ppb Na (Sodium)
≤5 ppb K (Potassium)

evapn. residue

≤2.0 mg/L

transmittance

210 nm, ≥60%
225 nm, ≥90%
245 nm, ≥98%

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a high purity solvent that ensures baseline stability, lowest impurity levels and high UV transmittance in LC-MS applications.

Application

Featured in screening and detection of estrogen-active nonylphenols in water by planar solid-phase extraction followed by high performance thin layer chromatography.
Also used in:
  • analysis of mineral oil saturated hydrocarbons in vegetable oils by LC-GC interface.[1]
  • studies on sugars and organic acids in biological matrices using GC coupled to triple quadruple mass spectrometry (QqQ-MS).[2]

Features and Benefits

  • no ghost peaks
  • full reproducibility
  • extends the lifetime of columns
  • optimized and tested for LC-MS applications
  • minimal background ion suppression
  • reduced risk of contamination
  • less exposure to hazardous chemicals

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.005 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Al (Aluminium): ≤ 10 ppb
Ca (Calcium): ≤ 10 ppb
Fe (Iron): ≤ 10 ppb
K (Potassium): ≤ 5 ppb
Mg (Magnesium): ≤ 10 ppb
Na (Sodium): ≤ 10 ppb
Transmission (at 210 nm): ≥ 60 %
Transmission (at 225 nm): ≥ 90 %
Transmission (from 245 nm): ≥ 98 %
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 2 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI negative): ≤ 20 ppb
Filtered by 0,2 µm filter.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A lab-developed interface for liquid-gas chromatography coupling based on the use of a modified programmed-temperature-vaporizing injector
Zoccali M, et al.
Journal of Chromatography A, 1622, 461096-461096 (2020)
Quantification of sugars and organic acids in biological matrices using GC-QqQ-MS
Chemosphere, 207-223 (2018)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service