Skip to Content
Merck

Skip To

UPS2

Proteomics Dynamic Range Standard Set

Protein Mass Spectrometry Calibration Standard

Synonym(s):

Dynamic Range Standards, Proteomics Standard Set

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12161503
EC Number:
232-650-8
NACRES:
NA.24
Form:
ready-to-use solution
Biological source:
human
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Level

form

ready-to-use solution

quality

Protein Mass Spectrometry Calibration Standard

concentration

10.6 μg/ampule protein

technique(s)

mass spectrometry (MS): suitable

shipped in

wet ice

storage temp.

−20°C

General description

The Proteomics Dynamic Range Standard Set is produced from a mixture of 47 individual human source or human sequence recombinant proteins, each of which has been selected to limit heterogeneous post-translational modifications (PTMs). The protein standard is formulated from 6 mixtures of 8 proteins to present a dynamic range of 5 orders of magnitude, ranging from 50 pmoles to 500 amoles. Each protein has been quantitated by amino acid analysis (AAA) prior to formulation.

Application

Proteomics Dynamic Range Standard Set has been used in the intensity-based absolute quantification (iBAQ) of E .coli proteins,[1] embryonic stem cells (ESCs) and neuronal precursor cells (NPCs) proteomes. It has also been used as a standard to spike HeLa cells for label-free quantification.
The Proteomics Dynamic Range Standard Set can be used to standardize and/or evaluate mass spectrometric (e.g., LC-MS/MS, MALDI-TOF-MS, etc.) and electrophoretic analysis conditions prior to the analysis of complex protein samples. UPS2 can be used to bracket precious experimental data sets between runs of a known complex standard sample. This allows confirmation of the robustness of the analysis method and stability of the instrument employed. Additionally, laboratories generating or comparing mass spectrometric data derived from poorly defined samples can use UPS2 as an external reference to assist with the evaluation of results and experimental methodology.
Proteomics Dynamic Range Standard Set has been used for the quantification of dynamic range universal protein standard on Orbitrap Analyzer using all ion fragmentation.[2] It has been used as a standard for intensity-based absolute quantification of proteins (iBAQ) in LC-MS (liquid chromatography-mass spectrometry)/MS analysis.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
UPS1EMS0004EMS0005
technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

-

technique(s)

-

biological source

human

biological source

-

biological source

-

biological source

-

concentration

10.6 μg/ampule protein

concentration

-

concentration

-

concentration

-

form

ready-to-use solution

form

ready-to-use solution

form

ready-to-use solution

form

ready-to-use solution

shipped in

wet ice

shipped in

-

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated 20 μg
    SDS

signalword

Danger

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



PANDA: A comprehensive and flexible tool for quantitative proteomics data analysis
Chang C, et al.
Bioinformatics, 35, 898-900 (2018)
Jacek R Wiśniewski et al.
Molecular systems biology, 8, 611-611 (2012-09-13)
We report a proteomic analysis of microdissected material from formalin-fixed and paraffin-embedded colorectal cancer, quantifying > 7500 proteins between patient matched normal mucosa, primary carcinoma, and nodal metastases. Expression levels of 1808 proteins changed significantly between normal and cancer tissues
Absolute proteome and phosphoproteome dynamics during the cell cycle of Schizosaccharomyces pombe (Fission Yeast).
Carpy A, et al.
Molecular and Cellular Proteomics, 13, 1925-1925 (2014)



Global Trade Item Number

SKUGTIN
UPS2-5X1SET04061837410352
UPS2-1SET04061837410345

Questions

  1. Regarding SKU# UPS2, can you please send me FASTA file of 48 proteins in UPS2 product? thanks you

    1 answer
    1. Helpful?

Reviews

No rating value

Active Filters