Skip to Content
Merck
All Photos(1)

Key Documents

46325

Supelco

Fomesafen

PESTANAL®, analytical standard

Synonym(s):

5-[2-Chloro-4-(trifluoromethyl)phenoxy]-N-(methylsulfonyl)-2-nitrobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClF3N2O6S
CAS Number:
Molecular Weight:
438.76
Beilstein:
8165046
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CS(=O)(=O)NC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C15H10ClF3N2O6S/c1-28(25,26)20-14(22)10-7-9(3-4-12(10)21(23)24)27-13-5-2-8(6-11(13)16)15(17,18)19/h2-7H,1H3,(H,20,22)

InChI key

BGZZWXTVIYUUEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Fomesafen may be used as a reference standard in the determination of fomesafen in soil samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengshan Yang et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 51(9), 1232-1239 (2011-12-01)
In order to find new strains to degrade fomesafen in contaminated soil, we isolated and identified a high-efficiency degrading bacterium from polluted soil. The degrading characteristics and remediation ability of the strain were also studied. Characteristics of morphological, physiological, biochemical
Qingming Zhang et al.
Environmental science and pollution research international, 20(1), 201-208 (2012-05-16)
Formesafen is a diphenyl ether herbicide that has adverse effects on non-target animals. However, knowledge about the effect of fomesafen on the antioxidant defense system in earthworms is vague. Thus, it is essential to investigate the effects of fomesafen on
Thierry Caquet et al.
Environmental toxicology and chemistry, 24(5), 1116-1124 (2005-08-23)
Ecotoxicological effects of the diphenyl ether herbicide fomesafen, applied alone or in combination with the adjuvant Agral 90 (mixture of polyethoxylated derivatives of nonylphenol), were assessed on planktonic communities in 18-m3 outdoor mesocosms during a nine-month study. Four mesocosms were
F Fakan et al.
Neoplasma, 45(2), 102-106 (1998-08-04)
Crystalline cytoplasmic needle-shaped inclusions in hepatocytes are considered to represent a specific morphological feature of porphyria cutanea tarda (PCT) and experimental PCT-like porphyrias. The cytoplasmic inclusions, however, are absent in hyperplastic hepatic nodules and hepatocellular carcinomas arising in the course
Marie-Agnès Coutellec et al.
Ecotoxicology (London, England), 15(2), 199-213 (2006-01-21)
Genetic and ecological factors may interact in their effects on fitness. Such interactions are thus to be expected between inbreeding and exposure of a population to a toxicant. The magnitude of inbreeding depression is thought to increase in stressful environments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service