Skip to Content
Merck
All Photos(2)

Documents

1.15333

Supelco

Water

suitable for LC/MS, LiChrosolv®

Synonym(s):

DI water, Deionized water, Distilled Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.03

product name

Water, for chromatography (LC-MS Grade) LiChrosolv®

vapor pressure

23 hPa ( 20 °C)

Quality Level

product line

LiChrosolv®

form

liquid

technique(s)

LC/MS: suitable

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

solubility

water: miscible

density

1.000 g/mL at 3.98 °C (lit.)

cation traces

Al: ≤10 ppb
Ca: ≤100 ppb
Fe: ≤5 ppb
K: ≤10 ppb
Mg: ≤20 ppb
Na: ≤20 ppb

application(s)

general analytical

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv® solvents have high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.

Application

Intended for U/HPLC and LC-MS applications.
Also featured in:
  • General methods for tests and analysis of food products
  • USP monograph methods for regulated pharmaceutical instrumental analysis
  • Method development for analysis of California list of pesticides in cannabis
  • Drug-protein binding measurements

Features and Benefits

  • Microfiltered by 0.2 µm filter
  • Packaged in glass bottles
  • Offers high resolution, sensitivity and optimized peak baseline separation
  • Provides rapid and reliable results in both electrospray ionization/atmospheric pressure chemical ionization (ESI/APCI) positive and negative ionization modes

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 5

1 of 5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service