Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 569,00 zł |
Informacje o tej pozycji
569,00 zł
material
stainless steel needle (point style 3)
needle size
22 ga (blunt tip)
feature
needle type SN (special needle)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80765
needle L
51 mm (2 in.)
volume
250 μL
compatibility
for sampling Valco VISF-2, for use with Rheodyne, for use with SSI
1 of 1
Ta pozycja | |||
|---|---|---|---|
| material stainless steel needle (point style 3) | material stainless steel needle (point style 3) | material stainless steel needle (point style 3) | material stainless steel needle (point style 2) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 80765 | manufacturer/tradename Hamilton 80665 | manufacturer/tradename Hamilton 80565 | manufacturer/tradename Hamilton 80384 |
| compatibility for sampling Valco VISF-2, for use with SSI, for use with Rheodyne | compatibility for use with Rheodyne, for use with SSI, for use with Valco VISF-2 | compatibility for use with Rheodyne, for use with SSI, mode of use Valco VISF-2 | compatibility for use with Hamilton 701SN (special needle) |
| needle size 22 ga (blunt tip) | needle size 22s ga (blunt tip) | needle size 22s ga (blunt tip) | needle size 26s ga (bevel tip) |
| volume 250 μL | volume 100 μL | volume 50 μL | volume 10 μL |
Still not finding the right product?
Explore all of our products under Hamilton® syringe, 700 series, special needle
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?