Documenti fondamentali
RAD196
radiello Outdoor Shelter
pk of 10
Scegli un formato
10 EA
341,00 €
341,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
10 EA
341,00 €
About This Item
Codice UNSPSC:
41104008
NACRES:
NB.24
341,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Confezionamento
pk of 10
Produttore/marchio commerciale
Radiello Code 196
tecniche
passive air sampling: suitable
applicazioni
air monitoring
environmental
industrial hygiene
Descrizione generale
A polypropylene protective outdoor shelter is available for housing up to four Radiello samplers during outdoor/ambient air sampling. The shelter allows for adequate ventilation while simultaneously protecting the samplers from harsh weather conditions. The shelter can be mounted to a variety of fixtures including lamp posts, traffic lights, and telephone poles of various diameters. The shelter can easily be tranported from the lab and mounted without the use of tools. Two mounting strips are included with each shelter.
Note legali
radiello is a trademark of Institi Clinici Scientifici Maugeri
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
Francesc A Esteve-Turrillas et al.
Journal of chromatography. A, 1216(48), 8549-8556 (2009-10-27)
A new procedure for the passive sampling in air of benzene, toluene, ethylbenzene and xylene isomers (BTEX) is proposed. A low-density polyethylene layflat tube filled with a mixture of solid phases provided a high versatility tool for the sampling of
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
P Lovreglio et al.
Giornale italiano di medicina del lavoro ed ergonomia, 33(2), 117-124 (2011-07-30)
To study the validity of urinary benzene as a biomarker of low and very low exposure to this toxicant, as compared with t,t-muconic acid (t,t-MA) and S-phenylmercapturic acid (SPMA), also taking into account the influence of cigarette smoking and co-exposure
Articoli
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocolli
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Contenuto correlato
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.