The dipper holds a volume of 12 mL. The overall length is 48cm and the diameter is 21mm. Please see the picture of the item attached.
Z150851
Liquid nitrogen dipper
for removing small quantities of liquid nitrogen (LN2) from storage dewars
Méret kiválasztása
78 290,00 Ft
Méret kiválasztása
About This Item
78 290,00 Ft
Javasolt termékek
Általános leírás
Compatible with the following range of cryogenic storage dewars, LD series: LD5, LD10, LD25, Classic 25, LD35 and LD50
Válasszon a legfrissebb verziók közül:
Analitikai tanúsítványok (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumentumok section.
-
What are the dimensions of this product?
1 answer -
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-