Merck
BR701352
BRAND® 96-well deep well plate, stackable
volume 1.1 mL, polystyrene
Synonym(e):
Deep-well Plates, Sample Storage
Größe auswählen
32 EA
€ 113,00
€ 113,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
32 EA
€ 113,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
€ 113,00
Check Cart for Availability
Materialien
polystyrene
Beschreibung
alphanumeric identification system
Sterilität
non-sterile
Leistungsmerkmale
skirt
Verpackung
pack of 32 ea
Hersteller/Markenname
BRAND 701352
Größe
96 well
Volumen
1.1 mL
Arbeitsvolumen
1.1 mL
Bindungsart
non-treated surface
Allgemeine Beschreibung
  • BRAND® 96-well deep well plates are made of high purity polystyrene (PS).
  • The ′U′ bottom ensures optimal sample mixture and collection.
  • Designed for a range of applications including High-Throughput Screening (HTS), serial dilutions, reagent transfer, and sample storage down to -80 °C.
  • Stackable for easy storage.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)