Fortfahren mit
Z379271
Deepwell 96 well plate
non-sterile
Synonym(e):
deepwell multiplate, storage plate, 378600000, F37860-0000, plate, untreated plate
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
24 ea
Warenkorb auf Verfügbarkeit prüfen
€ 315,00
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
clear polypropylene , round bottom clear wells (translucent)
Size:
96 wells
Sterility:
non-sterile
Binding type:
non-treated surface
€ 315,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear polypropylene , round bottom clear wells (translucent)
sterility
non-sterile
packaging
pack of 24 ea
manufacturer/tradename
Bel-Art F37860-0000
maximum volume
2 mL
size
96 wells
working volume
2 mL
binding type
non-treated surface
General description
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays, enzyme or receptor assays, or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.
Legal Information
Bel-Art is a registered trademark of Bel-Art Products, a division of SP
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z379298BAF378600001BR701342
description
non-sterile
description
Cover only
description
polypropylene, sterile, pack of 24 ea
description
volume 1.2 mL, polypropylene, elevated wells
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
binding type
non-treated surface
binding type
-
binding type
-
binding type
non-treated surface
manufacturer/tradename
Bel-Art F37860-0000
manufacturer/tradename
Bel-Art F37860-0004
manufacturer/tradename
Bel-Art F37860-0001
manufacturer/tradename
BRAND 701342
material
clear polypropylene , round bottom clear wells (translucent)
material
polypropylene
material
conical bottom clear polypropylene wells (translucent)
material
polypropylene , round bottom wells
working volume
2 mL
working volume
-
working volume
-
working volume
1.2 mL
size
96 wells
size
96 wells
size
96 wells
size
96 wells
Still not finding the right product?
Explore all of our products under Deepwell 96 well plate
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)