Merck
BR701358
BRAND® mat cover for deep well plate
volume 0.5 mL
Synonym(e):
Accessories, Cover mats, Deep-well Plates, Sample Storage
Größe auswählen
50 EA
€ 341,00
€ 341,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
€ 341,00
About This Item
UNSPSC-Code:
41122102
NACRES:
NB.22
€ 341,00
Check Cart for Availability
Sterilität
non-sterile
Hersteller/Markenname
BRAND 701358
Volumen
0.5 mL
Allgemeine Beschreibung
These cover mats reduce the maximum volume of wells but minimize evaporation.They can be used in conjunction with adhesive sealing films for added protection and containment.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)