Skip to Content
Merck
All Photos(1)

Key Documents

890890P

Avanti

18:1 TAP (DOTAP)

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-3-trimethylammonium-propane (chloride salt), DOTAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80NO4Cl
CAS Number:
Molecular Weight:
698.54
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g ((890890P-1G))
pkg of 2 × 100 mg ((890890P-200MG))
pkg of 5 × 100 mg ((890890P-100MG))
pkg of 1 × 25 mg ((890890P-25MG))

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Cl-]

InChI

1S/C42H80NO4.ClH/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-41(44)46-39-40(38-43(3,4)5)47-42(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2;/h20-23,40H,6-19,24-39H2,1-5H3;1H/q+1;/p-1/b22-20-,23-21-;

InChI key

KSXTUUUQYQYKCR-LQDDAWAPSA-M

Application

18:1 TAP (DOTAP) has been used in chloroform solutions to form dry film in the preparation of cationic liposomes. It has also been used in standard stock solution used for the preparation of cationic liposomes.

Biochem/physiol Actions

DOTAP (1,2-dioleoyl-3-trimethylammonium-propane) is a cationic lipid structure, used for in vitro and in vivo nucleic acid and protein delivery. The lipid composition and cell type affects the transfection potential of DOTAP. DOTAP mediates gene knockdown technique in hematopoietic stem cells (HSCs).

Packaging

5 mL Clear Glass Sealed Ampule (890890P-200mg)
5 mL Clear Glass Sealed Ampule (890890P-25mg)
5 mL Clear Glass Sealed Ampule (890890P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (890890P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of cationic liposomes by reversed-phase HPLC with evaporative light-scattering detection
Zhong Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 51(4), 947-951 (2010)
Efficient siRNA delivery by the cationic liposome DOTAP in human hematopoietic stem cells differentiating into dendritic cells
Martino S, et al.
BioMed Research International, 2009 (2009)
Cationic liposomes induce cytotoxicity in HepG2 via regulation of lipid metabolism based on whole-transcriptome sequencing analysis
Li Y, et al.
BMC Pharmacology & Toxicology, 19(1), 43-43 (2018)
DOTAP (and other cationic lipids): chemistry, biophysics, and transfection
Simberg D, et al.
Critical Reviews in Therapeutic Drug Carrier Systems, 21(4) (2004)
Elias J Sayour et al.
Nano letters, 18(10), 6195-6206 (2018-09-28)
Translation of nanoparticles (NPs) into human clinical trials for patients with refractory cancers has lagged due to unknown biologic reactivities of novel NP designs. To overcome these limitations, simple well-characterized mRNA lipid-NPs have been developed as cancer immunotherapeutic vaccines. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service