Skip to Content
Merck
All Photos(2)

Key Documents

P4417

Sigma-Aldrich

Phosphate buffered saline

1 of 3 reviewers received a sample product or took part in a promotion

tablet, pH 7.2-7.6 (1 tablet/200 mL)

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25
Pricing and availability is not currently available.

Product Name

Phosphate buffered saline, tablet

form

tablet

storage condition

dry at room temperature

color

white

pH

7.2-7.6 (1 tablet/200 mL)

solubility

water: soluble

density

1.02 g/cm3 at 20—25 °C
1.03 g/cm3 at 20—25 °C

application(s)

diagnostic assay manufacturing
histology
life science and biopharma

Looking for similar products? Visit Product Comparison Guide

General description

Phosphate-buffered saline (PBS) is a widely utilized buffer solution in biological research, valued for its versatility and essential attributes. Notably, its isotonicity closely mirrors the pH, osmolarity, and ion concentrations of the human body, rendering it non-toxic to cells. This pivotal characteristic plays a crucial role in diverse cell biology procedures, including the gentle washing of cells, safe transportation of tissues, and precise dilution of cells and reagents. These applications contribute to the preservation of cell viability and the establishment of a conducive physiological environment. The buffering capacity of PBS, sustained by a phosphate-phosphoric acid equilibrium, establishes a stable pH environment crucial for optimal enzyme activity and various biological processes. This makes PBS an ideal choice for tasks such as preparing reagents, protein dialysis, and immunohistochemistry, ensuring the reproducibility and reliability of experimental results.

Expanding beyond cell biology, PBS demonstrates versatility in applications such as creating custom ELISA buffers for optimal antibody-antigen interactions. Moreover, it serves as a proficient running buffer for Protein A/G chromatography, facilitating antibody purification based on specific antigen binding. In hematological and biochemical procedures, PBS plays a pivotal role in the effective washing of cells and tissues. In summary, PBS stands out as an indispensable tool in both biological and biochemical research, offering isotonicity, buffering capacity, simplicity, and stability. Its diverse applications in cell biology, reagent preparation, protein purification, and various biological assays underscore its profound significance in advancing scientific knowledge and understanding.

Application

Phosphate buffered saline has been used-
  • for the production of platinum nanoparticles (NP-Pt) hydrocolloids[1]
  • for the washing of blood sample and SEM (scanning electron microscope) stubs prior to SEM[1]
  • for preparing the solution of fluorescent lectin wheat germ agglutinin (WGA)[2]
  • for the dilution of capture antibody during ELISA (enzyme linked immunosorbent assay)[3]
  • for suspension of cell pellets during centrifugation[3]
  • in immunoassay procedures

Features and Benefits

  • High-quality product suitable for multiple research applications
  • Can be used as a buffer component in biological and biochemical applications
  • Highly soluble in water

Reconstitution

One tablet dissolved in 200 mL of deionized water yields 0.01 M phosphate buffer, 0.0027 M potassium chloride and 0.137 M sodium chloride, pH 7.4, at 25 °C.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Michaelis et al.
The journal of physical chemistry. B, 123(31), 6694-6704 (2019-07-11)
Although successful in the structural determination of ordered biomolecules, the spectroscopic investigation of oligopeptides in solution is hindered by their complex and rapidly changing conformational ensemble. The measured circular dichroism (CD) spectrum of an oligopeptide is an ensemble average over
Sputum cathelicidin, urokinase plasminogen activation system components, and cytokines discriminate cystic fibrosis, COPD, and asthma inflammation.
Xiao W et al
Chest, 128(4), 2316-2326 (2005)
Lipid rafts in the bovine milk fat globule membrane revealed by the lateral segregation of phospholipids and heterogeneous distribution of glycoproteins.
Lopez C et al
Food Chemistry, 120.1, 22-33 (2010)
Structural damage of chicken red blood cells exposed to platinum nanoparticles and cisplatin.
Kutwin M et al
Nanoscale Research Letters, 9(1), 257-257 (2014)
Simon C Drew et al.
PloS one, 5(12), e15875-e15875 (2011-01-07)
The amyloid-β (Aβ) peptide is the primary component of the extracellular senile plaques characteristic of Alzheimer's disease (AD). The metals hypothesis implicates redox-active copper ions in the pathogenesis of AD and the Cu(2+) coordination of various Aβ peptides has been

Protocols

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

See All

Questions

1–10 of 25 Questions  
  1. How much should one tablet weigh for it to yield 200ml 1xPBS? Asking just to make sure we won't use chipped tablets.

    1 answer
    1. Each tablet of this product should weigh between 1814.5 and 2005.5 milligrams.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. If I want to make the same buffer molarity as the product P4474-1L, how many tablets should I dissolve in 1L of DI water?

    1 answer
    1. P4417 and P4474 have different components and initial concentrations. One P4417 tablet dissolved in 200 mL of deionized water yields a 0.01 M phosphate buffer, 0.0027 M potassium chloride, and 0.137 M sodium chloride at pH 7.4, 25 °C. P4474 contains 1.058 mM potassium phosphate monobasic, 154 mM sodium chloride, and 2.97 mM sodium phosphate dibasic dihydrate. Due to these differences, it is not feasible to directly match the molarity of P4474 by dissolving P4417 tablets.

      Helpful?

  5. Going by instructions, 1 tab in 200 mL water gives a 1x PBS sol'n. The recipe I use for 1x PBS has the same salt concs. Sorry if this is a silly question, but can I make up a 3x PBS solution simply by dissolving 3 tablets in 200 mL of water?

    1 answer
    1. These tablets have not been tested for solubility or dissolution time at a 3X concentration. The end user would have to determine solubility experimentally. Note that 10X prepared solutions are available and may easily be diluted to the desired concentration. Please see the link below to review several of these product options:
      https://www.sigmaaldrich.com/search/pbs-10x?focus=products&page=1&perpage=30&sort=relevance&term=PBS%2010X&type=product

      Helpful?

  6. Hi! If with this product I want to do a 10x solution, do i dilute it in 2000 mL or first in 200mL and then do the dilution? Thanks!

    1 answer
    1. Dissolve 1 tablet in 200 ml of water to obtain: 0.01 M phosphate buffer, 0.0027 M potassium chloride and 0.137 M sodium chloride, pH 7.4 At 25 °C. These instructions are for a 1 X PBS preparation and this product is only intended for this specific concentration. If a 10X concentrate is needed, there are options in the catalog, such as the following products : 79383, P7059, 6506-OP, and 6505-OP.

      Helpful?

  7. Is it possible to obtain the salt components for product 6501-200TAB? Additionally, can you provide the composition details for other PBS tablets or powder products?

    1 answer
    1. The compositions are as follows:

      1. 6501-200TAB: This PBS contains 137mM NaCl, 2.7 mM KCl, and 10mM Na phosphate.

      2. P3813 - powder - Packed in Pouch or PAK: Each pouch contains 8 g Sodium Chloride, 0.2 g Potassium Phosphate, monobasic, 1.15 g Sodium Phosphate, dibasic, and 0.2 g Potassium Chloride, totaling 9.55 g per packet. This is a standard PBS formulation.

      3. P4417- Tablet: When dissolved in 200 mL of deionized water, one tablet yields 0.01 M phosphate buffer, 0.0027 M potassium chloride, and 0.137 M sodium chloride, at pH 7.4 and 25 °C. This product contains the same formulation as P3813 on a per liter basis (excluding tableting fillers): 8 g/L Sodium Chloride, 0.2 g/L Potassium Phosphate, monobasic, 1.15 g/L Sodium Phosphate, dibasic, and 0.2 g/L Potassium Chloride. This mixture should be diluted to one liter of water, resulting in a pH of 7.4, which can be adjusted using phosphoric acid or sodium hydroxide. The concentrations are 10 mM phosphate buffer, 2.7 mM potassium chloride, and 137 mM. These PBS tablets do not contain BSA.

      Helpful?

  8. What is the osmolality of P4417 made to 1X PBS at pH 7.4?

    1 answer
    1. This PBS once diluted in 200 mL distilled water will account for:
      2.7 mM potassium chloride = 5.4 mosmoles / L
      137 mM sodium chloride = 274 mosmoles / L

      The exact osmolality is not determined.

      Helpful?

  9. Is this PBS calcium-free?

    1 answer
    1. The formulation of this product is considered proprietary. However, calcium is not included in the composition.

      Helpful?

  10. What is the purity and CAS

    1 answer
    1. This product is a mixture of components. CAS numbers are not assigned to multi-component products. Likewise, as there are multiple components, purity is not reported. Upon reconstitution according to instructions, the solution will contain 0.01 M phosphate buffer, 0.0027 M potassium chloride, and 0.137 M sodium chloride.

      Helpful?

1–10 of 25 Questions  

Reviews

1 of 3 reviewers received a sample product or took part in a promotion
1–2 of 3 Reviews  

Active Filters

  1. Barcelona
    • Review 1
    • Vote 1
    5 out of 5 stars.

    Facilita la vida

    Durante mi tesis tenia que utilizar grandes cantidades y el poder hacerlo rapidamente con estas pastillas, te salva la vida

    Translate with Google

    Helpful?

  2. SETU, Kilkenny road , Carlow Ireland
    • Reviews 3
    • Votes 2
    5 out of 5 stars.

    good product

    arrived in time, very good product. I am happy with it

    Helpful?

  3. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service