Skip to Content
Merck
All Photos(1)

Key Documents

890880P

Avanti

18:0 TAP

Avanti Research - A Croda Brand

Synonym(s):

1,2-stearoyl-3-trimethylammonium-propane (chloride salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84NO4Cl
CAS Number:
Molecular Weight:
702.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 200 mg (890880P-200mg)
pkg of 1 × 25 mg (890880P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Cl-]

InChI key

NTTZBBIBMSBLNK-UHFFFAOYSA-M

Application

1,2-stearoyl-3-trimethylammonium-propane (18:0 TAP) might be used:
  • for the preparation of tri-loaded bilosomes (Tri-BLs) by thin film hydration technique
  • to formulate lipid nanoparticles (LNPs), for testing its efficiency of packaging mRNA into an LNP
  • to fabricate unilamellar empty liposomes
  • to prepare lipoplexes

Biochem/physiol Actions

1,2-stearoyl-3-trimethylammonium-propane (18:0 TAP) is a cationic lipid used in DNA transfection.

Packaging

5 mL Amber Glass Screw Cap Vial (890880P-200mg)
5 mL Amber Glass Screw Cap Vial (890880P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailing Yang et al.
Drug delivery, 26(1), 820-830 (2019-08-08)
Arthritis treatment has been challenging because of low drug exposure to the articular cavity. This study was intended to develop hyaluronic acid (HA)-functionalized bilosomes for targeted delivery of tripterine (Tri), an antiphlogistic phytomedicine, to the inflamed joint via ligand-receptor interaction.
Zhiqiang Sun et al.
Biomacromolecules, 13(10), 3395-3400 (2012-09-20)
Lipid-coated DNA nanoparticles (lipoplexes) are a powerful gene delivery tool with promising therapeutic applications. The mechanism of lipoplex assembly remains poorly understood. We explored DNA packing by a cationic lipid DSTAP (distearoyl trimethylammonium-propane) using magnetic tweezers. DSTAP-induced DNA condensation occurred
Maria Jose York-Duran et al.
Biomaterials science, 7(11), 4813-4826 (2019-09-20)
While ROS display crucial functions in many physiological processes, elevated ROS levels are also related to the initiation and progression of many severe diseases such as cancer, cardiovascular conditions or neurologic disorders. Research approaches to diminish ROS levels during disease
Siddharth Patel et al.
Journal of controlled release : official journal of the Controlled Release Society, 303, 91-100 (2019-04-16)
Retinal gene therapy has had unprecedented success in generating treatments that can halt vision loss. However, immunogenic response and long-term toxicity with the use of viral vectors remain a concern. Non-viral vectors are relatively non-immunogenic, scalable platforms that have had
Yuki Takechi-Haraya et al.
Langmuir : the ACS journal of surfaces and colloids, 34(26), 7805-7812 (2018-06-06)
It has recently been recognized that the mechanical properties of lipid nanoparticles play an important role during in vitro and in vivo behaviors such as cellular uptake, blood circulation, and biodistribution. However, there have been no quantitative investigations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service