Skip to Content
Merck
All Photos(2)

Key Documents

850375P

Avanti

18:1 (Δ9-Cis) PC (DOPC)

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; DOPC; PC(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1,2-dioleoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850375P-1g)
pkg of 2 × 100 mg (850375P-200mg)
pkg of 5 × 100 mg (850375P-500mg)
pkg of 1 × 25 mg (850375P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O

InChI

1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3/b22-20-,23-21-/t42-/m0/s1

InChI key

SNKAWJBJQDLSFF-AOMFJDGTSA-N

General description

18:1 (Δ9-Cis) PC or 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) is an unsaturated phospholipid.[1]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:1 (Δ9-Cis) PC (DOPC) has been used for reproducing the supramolecular structure of the synovial fluid.[2] It may also be used in the liposome preparation[3] and in the preparation of planar supported membranes.[4]

Biochem/physiol Actions

18:1 (Δ9-Cis) PC or 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) is mainly used in liposome preparation to deliver drug in to the body.[5] DOPC is used along with 1,2-dihexadecanoyl-sn-glycero-3-phosphocholine (DPPC) for lipid bilayer generation.[6]

Packaging

5 mL Clear Glass Sealed Ampule (850375P-200mg)
5 mL Clear Glass Sealed Ampule (850375P-25mg)
5 mL Clear Glass Sealed Ampule (850375P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (850375P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The size of lipid rafts: an atomic force microscopy study of ganglioside GM1 domains in sphingomyelin/DOPC/cholesterol membranes
Yuan C, et al.
Biophysical Journal, 82(5), 2526-2535 (2002)
Role of the biomolecular interactions in the structure and tribological properties of synovial fluid
Mirea D, et al.
Tribology International, 59(5), 302-311 (2013)
General model of phospholipid bilayers in fluid phase within the single chain mean field theory
Guo Y, et al.
J. Chem. Phys. , 140(17), 05B604-05B601 (2014)
?Force-from-lipids? gating of mechanosensitive channels modulated by PUFAs
Ridone P, et al.
Journal of the Mechanical Behavior of Biomedical Materials, 79(5), 158-167 (2018)
Formation of nanopores in DiynePC-DPPC complex lipid bilayers triggered by on-demand photo-polymerization
Chun MJ, et al.
Royal Society of Chemistry Advances, 8(49), 27988-27994 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service