Skip to Content
Merck
All Photos(3)

Documents

11836170001

Roche

cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail

Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial

Synonym(s):

protease inhibitor, protease inhibitor cocktail, roche protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

tablet

usage

sufficient for 25 10mL extraction solution preparations

packaging

vial of 25 tablets

manufacturer/tradename

Roche

technique(s)

inhibition assay: suitable

solubility

aqueous buffer: soluble

storage temp.

2-8°C

General description

cOmplete, Mini, EDTA-free Protease Inhibitor Tablets, inhibit a broad spectrum of serine and cysteine proteases. In contrast to other cOmplete tablets, they do not contain EDTA, leaving the stability and the function of metal-dependant proteins unaffected. The affinity purification of Poly-His-tagged fusion proteins using IMAC (Immobilized Metal Affinity Chromatography) is also facilitated (no dialysis necessary).
Due to the optimized composition of the tablets, they show excellent inhibition of serine and cysteine proteases, and well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete, Mini, EDTA-free tablets, contains both irreversible and reversible protease inhibitors. Metallo- and aspartic proteases are not inhibited.

cOmplete Mini, EDTA-free Tablets are identical to cOmplete Mini Tablets, the only difference being that no EDTA or other chelating (e.g. EGTA) agents are included.

Specificity

For the inhibition of serine, cysteine, but not metalloproteases.

Application

cOmplete, Mini, EDTA-free protease inhibitor cocktail has been used for the inhibition of serine and cysteine proteases in bacterial, yeast, plant, and animal cell extracts.
cOmplete, Mini, EDTA-free Tablets, are used for the inhibition of proteolytic activity in small volumes (up to 10 ml) in which EDTA may interfere with protein stability (e.g., metal-containing proteins) or subsequent assays. Since EDTA interferes with IMAC (Immobilized Metal Affinity Chromatography), cOmplete, Mini, EDTA-free, is preferentially used in the isolation process of Poly-His-tagged fusion proteins.
If it is necessary to inhibit proteolytic activity in a large volume (up to 50 ml), we recommend to use cOmplete, EDTA-free.
cOmpleteMini, EDTA-free Protease Inhibitor Cocktail has used as a component of
  • resuspension buffer for nuclear lysates preparation from prostate cancer cell lines for chromatin and RNA immunoprecipitation
  • resuspension buffer for protein extraction from neurospheres for western blot analysis
  • homogenization buffer for protein extraction from rat left ventricle samples and immunoprecipitaion buffer for human embryonic kidney 293 (HEK293) cells

Features and Benefits

Use cOmplete, Mini Protease Inhibitor Tablets to protect your proteins from a wide range of proteases. In just minutes, uninhibited proteolytic activity can degrade the protein you have spent days isolating. Each of these convenient water-soluble tablets contains a blend of protease inhibitors that inhibits proteolytic activity from most cell types, including animals, plants, yeast, and bacteria. No weighing or measuring is necessary, ensuring consistent results.

  • Easy-to-use: Simply drop a quick-dissolving tablet into your buffer.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.
  • Complete protection: Instantly protect your proteins against a broad range of proteases.
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.

Components

Proprietary mixture of several protease inhibitors.

Quality

Performance-tested with pancreas extract.

Physical form

Tablets in glass vials

Preparation Note

Working concentration: 1 tablet per 10 ml extraction solution
Working solution: Preparation of Working Solution:

One cOmplete Mini EDTA-free tablet is sufficient for the inhibition of the proteolytic activity in 10 ml extraction solution. When very high proteolytic activity is present, one tablet should be used for 7 ml extraction buffer. Tablets can be added directly to the extraction medium. Alternatively a stock solution (7 X conc.) can be prepared.

Stock solution (7 X conc.):

Dissolve one cOmplete Mini EDTA-free tablet in 1.5 ml dist. water or in 1.5 ml 100 mM phosphate buffer, pH 7.0.
Storage conditions (working solution): The stock solution can be stored at 2 to 8 °C for 1 to 2 weeks, or at least 12 weeks at -15 to -25 °C.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 7x stock solutions in 1.5 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1-2 weeks at 2 to 8 °C, or at least 12 weeks at -15 to -25 °C; can be used in thiol-containing solutions at 15 to 25 °C.

Legal Information

cOmplete is a trademark of Roche

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donald B Jump
Methods in molecular biology (Clifton, N.J.), 579, 375-389 (2009-09-19)
Very long chain fatty acids confer functional diversity on cells by variations in their chain length and degree of unsaturation. Microsomal fatty acid elongation represents the major pathway for determining the chain length of saturated, monounsaturated, and polyunsaturated fatty acids
Patrick R Punch et al.
Glycobiology, 30(7), 446-453 (2020-01-04)
High-dose irradiation poses extreme risk of mortality from acute damage to the hematopoietic compartment and gastrointestinal tract. While bone marrow transplantation can reestablish the hematopoietic compartment, a more imminent risk of death is posed by gastrointestinal acute radiation syndrome (GI-ARS)
Risk SNP-mediated promoter-enhancer switching drives prostate cancer through lncRNA PCAT19
Hua JT, et al.
Cell, 174(3), 564-575 (2018)
Andre Arashiro Pulschen et al.
Current biology : CB, 30(14), 2852-2859 (2020-06-06)
Live-cell imaging has revolutionized our understanding of dynamic cellular processes in bacteria and eukaryotes. Although similar techniques have been applied to the study of halophilic archaea [1-5], our ability to explore the cell biology of thermophilic archaea has been limited
Protein phosphatase 1c associated with the cardiac sodium calcium exchanger 1 regulates its activity by dephosphorylating serine 68-phosphorylated phospholemman
Hafver T, et al.
The Journal of biological chemistry, 291(9), 4561-4579 (2016)

Articles

Get maximum protection after protein isolation by using protease and phosphatase inhibitor cocktail tablets from Roche.

Protocols

Dissolve one tablet in 10 ml aqueous buffer or water. If high proteolytic activity is present, dissolve one tablet in 7 ml buffer.

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service