Skip to Content
Merck
All Photos(1)

Key Documents

B6916

Supelco

Bradford Reagent

for 0.1-1.4 mg/ml protein

Synonym(s):

Coomassie dye binding protein assay, Coomassie dye binding protein assay, Protein dye reagent, Protein dye reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161500
NACRES:
NA.32

Quality Level

form

solution

storage temp.

2-8°C

General description

Bradford assay is addition of coomassie brilliant blue G-250 to protein solution. The coomassie blue dye associates with basic and aromatic amino acids, thereby causing shift in absorbance during protein determination.

Application

Bradford Reagent has been used to determine total protein concentration.

Features and Benefits

  • The reagent is ready to use. No mixing or dilution required.
  • Color development is rapid. Only a five minute incubation and then the sample is read a 595 nm.
  • Reducing sugars and reducing substances along with thiols do not interfere with this reagent.
  • Reagent is suitable for micro (1-10 μg/ml) and standard (50-1400 μg/ml) assays.
  • Can be used in microwell plate assays.
  • Inexpensive assay.

Legal Information

Application

Product No.
Description
Pricing

also commonly purchased with this product

related product

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caleigh Mandel-Brehm et al.
Neurology, 93(5), e433-e444 (2019-07-05)
To identify molecular correlates of primary angiitis of the CNS (PACNS) through proteomic analysis of CSF from a biopsy-proven patient cohort. Using mass spectrometry, we quantitatively compared the CSF proteome of patients with biopsy-proven PACNS (n = 8) to CSF
Tracey Welham et al.
Journal of experimental botany, 60(12), 3353-3365 (2009-05-29)
Neutral/alkaline invertases are a subgroup, confined to plants and cyanobacteria, of a diverse family of enzymes. A family of seven closely-related genes, LjINV1-LjINV7, is described here and their expression in the model legume, Lotus japonicus, is examined. LjINV1 previously identified
Rosenberg IM
Protein Analysis and Purification: Benchtop Techniques (2006)
Proteomic response of the biological control fungus Trichoderma atroviride to growth on the cell walls of Rhizoctonia solani.
Grinyer J et al.
Current Genetics, 47, 381-381 (2005)
M Tal et al.
The Journal of biological chemistry, 260(18), 9976-9980 (1985-08-25)
Dimethyl sulfoxide was found to be effective for extraction of Coomassie Brilliant Blue R-250 (Coomassie R) from stained proteins on polyacrylamide gel slices. A good correlation was found between the ability of different proteins to bind Coomassie R and their

Protocols

To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.

To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.

Related Content

Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.

Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.

Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.

Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service