Skip to Content
Merck
All Photos(2)

Key Documents

850457P

Avanti

16:0-18:1 PC

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; POPC; PC(16:0/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850457P-1g)
pkg of 2 × 100 mg (850457P-200mg)
pkg of 5 × 100 mg (850457P-500mg)
pkg of 1 × 25 mg (850457P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-VYOBOKEXSA-N

General description

POPC is typically considered one of the model lipids for biophysical experiments. The fatty acid composition, i.e., saturated chain in the sn-1 position and unsaturated chain in the sn-2 position, mimics mammalian phospholipid composition. Since the major constituent in Egg PC is 1-palmitoyl-2-oleoyl PC, POPC is an excellent synthetic substitute for EPC.

Application

16:0-18:1 PC has been used in the preparation of liposomes for the muramyl tripeptide encapsulation.[1] It has also been used in the preparation of membrane scaffold protein 1 nanodisc.[2] 16:0-18:1 PC may be used in the synthesis of discoidal reconstituted high-density lipoproteins.[3]

Biochem/physiol Actions

1-palmitoyl-2-oleoyl-glycero-3-phosphocholine (POPC)or 16:0-18:1 PC plays an important role as a lung surfactant and reduces surface tension. POPC mimics natural cell membrane and may be useful for unsaturated lipids oxidation studies. Its interaction with ozone leads to oxidative decomposition at the oleoyl strand C-C moiety.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850457P-200mg)
5 mL Clear Glass Sealed Ampule (850457P-25mg)
5 mL Clear Glass Sealed Ampule (850457P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (850457P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorophore Labeling, Nanodisc Reconstitution and Single-molecule Observation of a G Protein-coupled Receptor
Lamichhane R, et al.
Bio-protocol, 7(12) (2017)
Tripta Bhatia et al.
ACS nano, 12(5), 4478-4485 (2018-04-17)
Giant unilamellar vesicles (GUVs) provide a direct connection between the nano- and the microregime. On the one hand, these vesicles represent biomimetic compartments with linear dimensions of many micrometers. On the other hand, the vesicle walls are provided by single
Oxidative degradation of the monolayer of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) in low-level ozone
Qiao L, et al.
The Journal of Physical Chemistry B, 119(44), 14188-14199 (2015)
Modified apolipoprotein (apo) AI by artificial sweetener causes severe premature cellular senescence and atherosclerosis with impairment of functional and structural properties of apoA-I in lipid-free and lipid-bound state
Jang W, et al.
Molecules and Cells, 31(5), 461-470 (2011)
Bianca Buchegger et al.
Analytical chemistry, 90(21), 12372-12376 (2018-10-24)
Mobility of proteins and lipids plays a major role in physiological processes. Platforms which were developed to study protein interaction between immobilized and mobile proteins suffer from shortcomings such as fluorescence quenching or complicated fabrication methods. Here we report a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service