Skip to Content
Merck
All Photos(1)

Key Documents

D6429

Sigma-Aldrich

DMEM - high glucose

Dulbecco’s Modified Eagle’s Medium - high glucose
2 of 6 reviewers received a sample product or took part in a promotion

With sodium bicarbonate, ʟ-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

Product Name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

HEPES: no
NaHCO3: yes
phenol red: yes
sodium pyruvate: yes
L-glutamine: yes
glucose: high

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This DMEM-Hi glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".
Dulbecco′s Modified Eagle′s Medium - high glucose has been used for cell culture.[1][2][3]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouse sphingosine kinase isoforms SPHK1a and SPHK1b differ in enzymatic traits including stability, localization, modification, and oligomerization.
Kihara A, et al.
The Journal of Biological Chemistry, 281(7), 4532-4539 (2006)
Kazuhiro Ikeda et al.
Scientific reports, 7(1), 2850-2850 (2017-06-08)
Human pluripotent stem cells are a potentially powerful cellular resource for application in regenerative medicine. Because such applications require large numbers of human pluripotent stem cell-derived cells, a scalable culture system of human pluripotent stem cell needs to be developed.
Neuraminidase receptor binding variants of human influenza A(H3N2) viruses resulting from substitution of aspartic acid 151 in the catalytic site: a role in virus attachment?
Lin YP, et al.
Journal of Virology, 84(13), 6769-6781 (2010)
Kenta Imai et al.
Journal of cell science, 129(20), 3781-3791 (2016-09-03)
Autophagy is an intracellular degradation pathway conserved in eukaryotes. Among core autophagy-related (Atg) proteins, mammalian Atg9A is the sole multi-spanning transmembrane protein, and both of its N- and C-terminal domains are exposed to the cytoplasm. It is known that Atg9A
Mikiko Fukuda et al.
The Journal of reproduction and development, 62(1), 121-125 (2015-11-26)
Production of knockout mice using targeted embryonic stem cells (ESCs) is a powerful approach for investigating the function of specific genes in vivo. Although the protocol for gene targeting via homologous recombination (HR) in ESCs is already well established, the

Articles

Cell culture protocol for passaging and splitting suspension cell lines

This discussion will highlight some of these methodologies, namely, the use of Multiple Reaction Monitoring (MRM) and Protein-AQUA.

Related Content

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Questions

1–2 of 2 Questions  
  1. After opening a bottle of medium: how long can I use this bottle ?(stored at 4C°)

    1 answer
    1. Provided that the product is handled and stored properly and has not been contaminated, the medium will be stable up to the expiration date. This date is provided on the label and the lot-specific Certificate of Analysis.

      Helpful?

  2. I would like to inquire about the amino acid formulation in DMEM (#6429, #6546). Specifically, could you provide information on the concentration of aspartate and glutamate in these formulations? Thank you very much.

    1 answer
    1. The composition of this product is published on page 3 of the product information sheet. Please see the link below to access this document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/318/256/d6429for.pdf

      Helpful?

Reviews

2 of 6 reviewers received a sample product or took part in a promotion

Active Filters

  1. Atlanta, GA
    • Reviews 4
    • Votes 9
    5 out of 5 stars.

    D6171 with HEPES modification

    The best media to culture HEK293FT cells. The HEPES modification seems beneficial during calcium-phosphate transfection by perhaps stabilizing pH.

    Helpful?

  2. 5 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service