Skip to Content
Merck
All Photos(1)

Documents

D5671

Sigma-Aldrich

DMEM - high glucose

With sodium Bicarbonate, without ʟ-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
glucose: high
NaHCO3: yes
sodium pyruvate: no
phenol red: yes
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This DMEM-Hi glucose medium is a 1x complete medium with no added factors (common modifications) such as L-alanyl-L-glutamine, HEPES, or sodium pyruvate. It differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Reconstitution

Supplement with 0.584 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smita Krishnaswamy et al.
PloS one, 13(10), e0203389-e0203389 (2018-10-30)
Cellular regulatory networks are not static, but continuously reconfigure in response to stimuli via alterations in protein abundance and confirmation. However, typical computational approaches treat them as static interaction networks derived from a single time point. Here, we provide methods
Massiel Cepeda-Molero et al.
PLoS pathogens, 13(10), e1006706-e1006706 (2017-10-31)
Enteropathogenic E. coli (EPEC) is a human pathogen that causes acute and chronic pediatric diarrhea. The hallmark of EPEC infection is the formation of attaching and effacing (A/E) lesions in the intestinal epithelium. Formation of A/E lesions is mediated by
Marwa El Soury et al.
Frontiers in molecular neuroscience, 11, 157-157 (2018-06-06)
Peripheral nerves are characterised by the ability to regenerate after injury. Schwann cell activity is fundamental for all steps of peripheral nerve regeneration: immediately after injury they de-differentiate, remove myelin debris, proliferate and repopulate the injured nerve. Soluble Neuregulin1 (NRG1)
Victoria Gillan et al.
Frontiers in cellular and infection microbiology, 7, 452-452 (2017-12-07)
Resistance to anthelmintic drugs is a major problem in the global fight against parasitic nematodes infecting humans and animals. While previous studies have identified mutations in drug target genes in resistant parasites, changes in the expression levels of both targets
Zay Yar Oo et al.
Molecular oncology, 13(7), 1503-1518 (2019-05-03)
Drugs such as gemcitabine that increase replication stress are effective chemotherapeutics in a range of cancer settings. These drugs effectively block replication and promote DNA damage, triggering a cell cycle checkpoint response through the ATR-CHK1 pathway. Inhibiting this signalling pathway

Articles

High titer lentiviral particles for LC3 variants used for live cell analysis of cellular autophagy.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

Protocols

This detailed procedure allows you to transduce Mouse Embryonic Fibroblasts (MEF) using MISSION ExpressMag Super Magnetic Kit.

XTT assay protocol for measuring cell viability, proliferation, activation and cytotoxicity. Instructions for XTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service