Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
235-747-3
MDL number:
Specific activity:
≥39,000 units/mg protein
Biological source:
chicken egg white
biological source
chicken egg white
Quality Segment
form
powder or granules
specific activity
≥39,000 units/mg protein
mol wt
single-chain 14.3 kDa
technique(s)
cell based assay: suitable
suitability
suitable for cell lysis
UniProt accession no.
application(s)
cell analysis
storage temp.
room temp
Gene Information
chicken ... LYZ(396218)
Application
Lysozyme from chicken egg white has been used for the extraction of genomic DNA from bacterial cells. It has been used as an external standard for MALDI-TOF (matrix assisted laser desorption ionization-time of flight) mass analysis.
Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.
Biochem/physiol Actions
Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan. Gram-negative bacteria are less susceptible due to the presence of an outer membrane and a lower proportion of peptidoglycan. However, these cells may be hydrolyzed in the presence of EDTA that chelates metal ions in the outer bacterial membrane.
The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).
The enzyme is active over a broad pH range (6.0 to 9.0). At pH 6.2, maximal activity is observed over a wider range of ionic strengths (0.02 to 0.100 M) than at pH 9.2 (0.01 to 0.06 M).
Lysozyme hydrolyzes β(1→4) linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrin. Gram-positive cells are quite susceptible to this hydrolysis as their cell walls have a high proportion of peptidoglycan.
Features and Benefits
Lysozyme from chicken egg white is a versatile enzyme with unique properties that make it effective for various applications, including bacterial cell wall breakdown, genomic DNA extraction, and external standard for mass analysis. It is reliable, active over a broad pH range, and available in various packaging sizes.
Other Notes
One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.
View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer
Still not finding the right product?
Explore all of our products under Lysozyme from chicken egg white
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| L6876-5G | 04061835573523 |
| L6876-25G | 04061835516148 |
| L6876-10G | 04061835521920 |
| L6876-1G | 04061835573516 |
| L6876-100G | 04061835512706 |
