Kluczowe dokumenty
Z645745
Mini chemical spill kit
Wybierz wielkość
1 KIT
228,00 zł
228,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Wybierz wielkość
Zmień widok
1 KIT
228,00 zł
About This Item
Kod UNSPSC:
47131905
NACRES:
NB.35
228,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
producent / nazwa handlowa
Clear-spill CK1
Opis ogólny
Quick response kit designed for smaller spills. Contains 10 pads and 2 x 8 cm by 1.2 m socks supplied in neat bag measuring approx. 50 cm x 50 cm x 10 cm with instructions. Kit will absorb 13 litres.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Y Y Xiao et al.
Epidemiology and infection, 143(9), 1833-1838 (2014-11-02)
No published studies have discussed details of the prognosis and survival of patients with severe avian influenza A(H7N9) infection. In this study we analysed 128 laboratory-confirmed cases of severe H7N9 infection in Zhejiang province, the most affected region during the
Giovanna Trinchese et al.
The Journal of nutritional biochemistry, 26(11), 1136-1146 (2015-06-30)
Different nutritional components are able, by modulating mitochondrial function and gut microbiota composition, to influence body composition, metabolic homeostasis and inflammatory state. In this study, we aimed to evaluate the effects produced by the supplementation of different milks on energy
Mark Zeller et al.
Molecular biology and evolution, 32(8), 2060-2071 (2015-04-11)
The majority of human group A rotaviruses possess the P[8] VP4 genotype. Recently, a genetically distinct subtype of the P[8] genotype, also known as OP354-like P[8] or lineage P[8]-4, emerged in several countries. However, it is unclear for how long
Monique Silvy et al.
Transfusion, 55(6 Pt 2), 1407-1410 (2014-11-22)
The rare amorph Rhnull phenotype is caused by silent alleles at the RH locus and usually arises in consanguineous families. To date, only five molecular backgrounds have been identified in five unrelated families. Subjects with Rhnull red blood cells (RBCs)
James Neil Fisher et al.
Oncotarget, 6(15), 13176-13200 (2015-05-12)
SKBR3-cells, characterized by ERBB2/RARA co-amplification, represent a subgroup of HER2+ breast-cancers sensitive to all-trans retinoic acid (ATRA) and Lapatinib. In this model, the two agents alone or in combination modulate the expression of 174 microRNAs (miRs). These miRs and predicted