Merck
CLS4923
Corning® pipetting aids
Air filter replacement
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
メーカー/製品名
Corning 4923
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Costar Stripettors are lightweight, adjustable-speed pipetting aids designed for use with all serological pipettes. The nose cone of the units are autoclavable and fitted with a replaceable 0.2 μm hydrophobic sterilizing filter for added protection against biohazardous material. The Stripettor operates on a rechargeable 9V NiCd battery and features an LED light on the handle that lets the user know when to recharge. The unit is fully operational while recharging.
法的情報
Corning is a registered trademark of Corning, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)