Merck
513010
Miniature Regulator with Gauge
About This Item
UNSPSC kód:
41112500
Általános leírás
Reliable pressure regulation to 1 psig (0.07 kg/cm2), indicated on a 0-60 psi (4.2 kg/cm2) gauge. Easily connects to SCOTTY 14 cylinders with the 1/8-inch NPT connector provided. You can also attach the syringe adapter (Cat. No. 609010) to the regulator for low pressure sample removal. Aluminum body with acetyl resin bonnet. Maximum inlet pressure: 400 psi (28.1 kg/cm2).
Vigyázat
Cannot be used with Oxygen
Válasszon a legfrissebb verziók közül:
Analitikai tanúsítványok (COA)