Z114235
Hand-operated aluminum cap crimper
O.D. 13 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Doporučené produkty
O.D.
13 mm
General description
- Used to attach aluminum seals to bottles and vials with a crimp/serum finish
- Cushioned ergonomic handle reduces hand fatigue
- Labeled for quick size identification
- Polished crimping jaws provide consistent sealing
- 2 year warranty
related product
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)