Merck
72200
Sigma-Aldrich
Neufuchsin
for microscopy
Synonym(e):
Basic Violet 2, Fuchsin N
About This Item
Lineare Formel:
C22H24N3Cl
CAS-Nummer:
Molekulargewicht:
365.90
Farbindexnummer:
42520
Beilstein:
8740867
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352200
PubChem Substanz-ID:
NACRES:
NA.25
Qualität
for microscopy
Form
solid
λmax
553 nm
SMILES String
[Cl-].Cc1cc(ccc1N)C(\c2ccc(N)c(C)c2)=C3/C=CC(=[NH2+])C(C)=C3
InChI
1S/C22H23N3.ClH/c1-13-10-16(4-7-19(13)23)22(17-5-8-20(24)14(2)11-17)18-6-9-21(25)15(3)12-18;/h4-12,23H,24-25H2,1-3H3;1H
InChIKey
IPSIPYMEZZPCPY-UHFFFAOYSA-N
Rechtliche Hinweise
Magenta is a trademark of Merck KGaA, Darmstadt, Germany
Signalwort
Warning
Gefahreneinstufungen
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Zielorgane
Respiratory system
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
J Kriegsmann et al.
Laboratory investigation; a journal of technical methods and pathology, 71(6), 911-917 (1994-12-01)
A reliable, sensitive, and specific double labeling technique is required that allows the simultaneous visualization of in situ hybridization products and antigens. Currently used double labeling techniques are limited by various problems including the numerous disadvantages associated with radioactive labels
G B Proctor et al.
Stain technology, 63(2), 101-111 (1988-03-01)
Chemical properties of Weigert's resorcin-fuchsin, orcinol-new fuchsin, Sheridan's crystal violet and their resorcinol-free analogues were investigated using reverse-phase and gel filtration chromatography, electrophoresis, and visible light spectroscopy. Their staining properties were also studied. It was concluded that 1) the staining
M Fujita et al.
[Hokkaido igaku zasshi] The Hokkaido journal of medical science, 60(3), 313-320 (1985-05-01)
In the early stage of myocardial infarction, the mortality rate is very high and its diagnosis is difficult not only clinically but also pathologically. Hematoxylin basic fuchsin picric acid (HBFP) staining method has been controversial on reliability for the morphologic
T E Sehlinger et al.
Stain technology, 62(5), 291-297 (1987-09-01)
Commercial samples of basic fuchsin contain variable proportions of four homologs, pararosaniline, rosaniline, magenta II, and new fuchsin. That different samples of dyes give variable staining is documented in the literature. Three commercial samples of basic fuchsin were investigated using
Dye standards, Part II. 8: New fuchsin (CI 42520). European Committee for Clinical Laboratory Standards (ECCLS), Subcommittee on Reference Materials for Tissue Stains (SRMTS).
The Histochemical journal, 24(4), 240-242 (1992-04-01)