Kluczowe dokumenty
Z655422
Hanna rugged platinium & gold tips ORP electrodes for general use
Gold pin tip - reduction reactions
About This Item
Kod UNSPSC:
41115603
producent / nazwa handlowa
Hanna HI 4430B
Parametry
0-80 °C temp. range
2 bar max. pressure
Opis ogólny
  • Single, Ag/AgCI
  • Ceramic, single junction
  • Electrolyte - gel
  • 1 m coaxial cable
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)