安全性情報
21532
SGE社ガスタイトシリンジ, ルアーロック型
50MR-LL-GT, volume 50 mL, needle size (not included)
別名:
シリンジ, SGE社製, 50MAX-LL-GT, 50 mL
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥54,500
¥54,500
Check Cart for Availability
バルクの問い合わせ
サイズを選択してください
表示を変更する
1 EA
¥54,500
About This Item
UNSPSCコード:
41122001
¥54,500
Check Cart for Availability
バルクの問い合わせ
物質
glass syringe
ニードルサイズ
(not included)
特徴
needle type Luer lock
包装
pkg of 1 ea
メーカー/製品名
SGE 009660
容量
50 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
  • SGE gas tight syringe without a needle, Model: 50MR-LL-GT.
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel, which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
SGE gas tight syringes are constructed of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones where applicable. An interference fit between the plunger tip and the barrel enhances the gas sealing properties of the syringe. Plungers are interchangeable. Syringes can be autoclaved, but the plunger should be removed.
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
Mark Trimmer et al.
Applied and environmental microbiology, 69(11), 6447-6454 (2003-11-07)
Until recently, denitrification was thought to be the only significant pathway for N(2) formation and, in turn, the removal of nitrogen in aquatic sediments. The discovery of anaerobic ammonium oxidation in the laboratory suggested that alternative metabolisms might be present
Widespread occurrence of the anammox reaction in estuarine sediments.
Nicholls JC and Trimmer M.
Aquatic Toxicology (Amsterdam, Netherlands), 55 (2), 105-105 (2009)
Mark Henning et al.
Rapid communications in mass spectrometry : RCM, 21(14), 2269-2272 (2007-06-20)
Compound-specific deltaD and delta13C analyses of gas mixtures are useful indicators of geochemical and environmental factors. However, the relative concentrations of individual components in gas mixtures (e.g., H2, CO2, methane, ethane, propane, i-butane, n-butane) may vary over several orders of