Documenti fondamentali
22731
Supelco
SS-174 PTFE-Faced Septa
diam. 11 mm ( 7/16 in.)
Scegli un formato
100 EA
220,00 €
220,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
100 EA
220,00 €
About This Item
Codice UNSPSC:
41115718
220,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Materiali
off-white silicone body (123 mil thick)
yellow PTFE face (2 mil thick)
Confezionamento
pkg of 100 ea
Diametro
11 mm ( 7/16 in.)
Descrizione generale
PTFE-faced septa are designed for applications where greater inertness is required. The off-white silicone rubber body (123 mil thick) provides plenty of support for the yellow PTFE face (2 mil thick). After the PTFE is ruptured during injection, some bleeding may occur. Because this bleed increases as more of the rubber is exposed, we recommend using a needle guide to reduce the size of the punctured area. For use with 200 °C to 300 °C inlet temperatures.
PTFE-fused septa are suitable to overcome the degradation due to prolonged high temperatures which occurs in silicone rubber septa. The degradation results in baseline drift and additional peaks. It is generally fixed with the PTFE face towards the column packing.[1]
Applicazioni
Glass vials equipped with stir bars may be sealed with PTFE faced silicone septa for solvent drop microextraction analysis of water samples.[2]
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
Carmen López-Blanco et al.
Analytical and bioanalytical chemistry, 383(4), 557-561 (2005-09-01)
Water contamination due to the wide variety of pesticides used in agriculture practices is a global environmental pollution problem. Analytical methods with low quantification limits are necessary. The application of a new extraction technique, solvent drop microextraction (SDME), followed by
Ian A. Fowlis
Gas Chromatography, 62-62 (2008)