Fontos dokumentumok
EP5910751001
Adapter for Eppendorf® S-4x Universal Large Rotor
for 50 mL conical tubes/microtiter plates/250 mL flat-bottom or 175-225 mL conical tubes/microtiter plates, pkg of 2 ea
About This Item
UNSPSC kód:
41103911
NACRES:
NB.82
Jellemzők
Holds 250 mL flat bottom bottles
for 50 mL conical tubes/microtiter plates/250 mL flat-bottom or 175-225 mL conical tubes/microtiter plates
kiszerelés
pkg of 2 ea
gyártó/kereskedő neve
Eppendorf® 5910751001
technika/technikák
centrifugation: suitable
kompatibilitás
for use with Universal bucket
Általános leírás
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Jogi információk
Eppendorf is a registered trademark of Eppendorf AG