Hlavní dokumenty
Z509035
Sigma-Aldrich
Pipette bulb
volume 1 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
natural rubber
thimble blue
packaging
pkg of 24 ea
manufacturer/tradename
200-1C
volume
1 mL
General description
Natural rubber dropper bulbs for use with pipettes ensuring precise transfer and measurement of liquids.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Slide 1 of 10
1 of 10
Pipette bulb capacity 35 mL
Z186694
Pipette bulb
Pipette fillers red natural rubber bulb type
Z136468
Pipette fillers
Atomizer bulb useful for priming liquid-transfer lines
Z186724
Atomizer bulb
Pipette bulb capacity 60 mL
Z136069
Pipette bulb
Pipette bulb capacity 150 mL
Z186708
Pipette bulb
Pipette fillers red silicone bulb type
Z239089
Pipette fillers
Squeeze-bulb pump internal flapper valve prevents reverse flow, pkg of 1 ea
Z200336
Squeeze-bulb pump