Key Documents
Z136468
Pipette fillers
red natural rubber bulb type
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
red natural rubber bulb
manufacturer/tradename
RED-001
General description
Valved, choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.