Key Documents
Z186708
Pipette bulb
capacity 150 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
natural rubber
oval red
packaging
pkg of 3 ea
manufacturer/tradename
200-150
capacity
150 mL
Zákazníci si také prohlíželi
Slide 1 of 6
1 of 6
Pipette fillers red natural rubber bulb type
Z136468
Pipette fillers
Bel-Bulb™ pipettor vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm
Z136085
Bel-Bulb pipettor
Atomizer bulb useful for priming liquid-transfer lines
Z186724
Atomizer bulb
Pipette bulb capacity 5 mL
Z186678
Pipette bulb