Merck
CAD6020
1-way stopcock
female Luer lock to female Luer lock
Größe auswählen
1 EA
CHF 110.00
CHF 110.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 110.00
About This Item
UNSPSC-Code:
41122000
NACRES:
NB.03
CHF 110.00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Cadence Science 6020
Verwandte Kategorien
Allgemeine Beschreibung
Stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)