Merck
CAD6001
3-way stopcock
female Luer lock to male Luer, right side, female Luer lock, compression-nut
Größe auswählen
1 EA
CHF 155.00
CHF 155.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 155.00
About This Item
UNSPSC-Code:
41122000
NACRES:
NB.03
CHF 155.00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Cadence Science 6001
Verwandte Kategorien
Allgemeine Beschreibung
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)