Merck
CAD6017
3-way stopcock
female Luer lock to male Luer lock, left side, female Luer lock
Größe auswählen
1 EA
CHF 194.00
CHF 194.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 194.00
About This Item
UNSPSC-Code:
41122000
NACRES:
NB.03
CHF 194.00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Cadence Science 6017
Verwandte Kategorien
Allgemeine Beschreibung
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Can you autoclave this 3-way port? Can you use it to hook up on a sterile fermentation circuit?
    1 answer
    1. This product is supplied by Cadence Science. The manufacturer has not determined autoclavability for non-medical devices, such as this item. In general, chrome plated brass may be autoclaved, however brass is subject to corrosion. Other sources suggest an extended dry time when autoclaving. The suitability for use for fermentation systems would have to be determined experimentally by the end user.