Merck
HPLC analysis of Methyl Paraben and Organic Acids on Ascentis® Express RP-Amide
HPLC analysis of Methyl Paraben and Organic Acids on Ascentis® Express RP-Amide application for HPLC
Materiais
coluna analítica
Nº do produto
Descrição
Preços
Ascentis® Express RP-Amide, 5 μm HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
padrão
Nº do produto
Descrição
Preços
Ácido acetilsalicílico
analytical standard
Uracil
≥99.0%
Dehydroacetic acid
≥98.0% (T)
3-Fluorobenzoic acid
97%
Benzoic acid
analytical standard
Metil 4-hidroxibenzoato
PESTANAL®, analytical standard
Ácido 4-aminobenzoico
analytical standard
CONDITIONS
column
Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (50774-U)
mobile phase
[A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate
1.3 mL/min
pressure
1870 psi (129 bar)
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
[A] water; [B] acetonitrile; (50:50, A:B)